Learn More
Abnova™ ABCC11 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00085320T01
Catalog No. 89-932-122
Add to cart
Description
Description
Specifications
Specifications
293T
BC039085
pCMV-ABCC11 full-length
100 μL
Human
ABCC11 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 62.1kDa
Transient overexpression cell lysate was tested with anti-ABCC11 antibody (Mfr. No. H00085320-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot