Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548527

Catalog No. PA548527

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Probable transporter.
Specifications

Specifications

MRP9
Polyclonal
PBS with 0.09% sodium azide
Q96J65, Q80WJ6
Multidrug resistance-associated protein 9, ATP-binding cassette sub-family C member 12, ABCC12, MRP9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
244562, 94160
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ABCC12
Liquid
ABCC12
KLH conjugated synthetic peptide between 723-752 amino acids from the Central region of human ABCC12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit