Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCC12 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169063
Description
ABCC12 Polyclonal specifically detects ABCC12 in Mouse samples. It is validated for Western Blot.Specifications
ABCC12 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q80WJ6 | |
Abcc12 | |
Synthetic peptides corresponding to Abcc12 (ATP-binding cassette, sub-family C (CFTR/MRP), member 12) The peptide sequence was selected from the middle region of Abcc12. Peptide sequence NILFGEKYNHQRYQHTVHVCGLQKDLNSLPYGDLTEIGERGVNLSGGQRQ. | |
Affinity Purified | |
RUO | |
94160 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ATP-binding cassette transporter sub-family C member 12, ATP-binding cassette, sub-family C (CFTR/MRP), member 12, MGC27071, MRP9ATP-binding cassette sub-family C member 12, multidrug resistance-associated protein 9 | |
Rabbit | |
153 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title