Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC5 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271540AF647

Catalog No. N271540A647


Add to cart

Description

Description

ABCC5 Monoclonal antibody specifically detects ABCC5 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ABCC5
OTI2C8
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ABCC5
Full length recombinant protein of human ABCC5 (NP_001018881) produced in E.coli.
Immunogen affinity purified
ABC Transporters, Amino Acids Drugs and other small molecules, Lipid and Metabolism
Primary
10057.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ATP-binding cassette sub-family C member 5, ATP-binding cassette, sub-family C (CFTR/MRP), member 5, canalicular multispecific organic anion transporter C, EST277145, MOAT-CABC33, MRP5MOATC, multidrug resistance-associated protein 5, Multi-specific organic anion transporter C, pABC11, SMRPDKFZp686C1782
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit