Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcc8/Abcc9 Mouse anti-Human, Rat, APC, Clone:S323A-31, Abnova™

Mouse monoclonal antibody raised against partial recombinant rat Abcc8/Abcc9.

Manufacturer:  Abnova Corporation MAB17053

Catalog No. 89-333-821


Add to cart

Specifications

Specifications

Abcc8/Abcc9
S323A-31
ATP-binding cassette, sub-family C (CFTR/MRP), member 8
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Liquid
Mouse
IgG1
Store at -20°C.
Monoclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
APC
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Abcc8
Sur, Sur1
Recombinant protein corresponding to amino acids 1503-1545 at C-terminus of rat Abcc9.
100μg
Antibody
25559, 25560
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit