Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcc8, Mouse anti-Rat, ATTO655, Clone: S289-16, Abnova
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18327
Specifications
Abcc8 | |
S289-16 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
ATP-binding cassette, sub-family C (CFTR/MRP), member 8 | |
Sur|Sur1 | |
Mouse | |
IgG1 | |
RUO | |
Antibody | |
Monoclonal | |
Rat |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
ATTO655 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Liquid | |
Abcc8 | |
Recombinant protein corresponding to amino acids 1548-1582 at C-terminus of rat Abcc8. | |
100 μg | |
Store at -20°C. | |
Primary | |
25559 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title