Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567620
Description
Predicted to react with Mouse samples.
This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
ABCD1 | |
Polyclonal | |
Unconjugated | |
ABCD1 | |
ABC42; ABCD1; Adrenoleukodystrophy protein; ALD; Aldgh; ALDP; AMN; ATP binding cassette subfamily D member 1; ATP-binding cassette sub-family D member 1; ATP-binding cassette, subfamily D (ALD), member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; RGD1562128; X-linked adrenoleukodystrophy (ALD) gene homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11666, 215 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P33897, P48410 | |
ABCD1 | |
Synthesized peptide derived from human ABCD1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction