Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522246

Catalog No. PIPA522246


Only null left
Add to Cart

Description

Description

Recommended positive controls: SHSY5Y. Predicted reactivity: Mouse (94%), Rat (92%), Chicken (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCD2
Polyclonal
Unconjugated
ABCD2
ABC39; Abcd2; adrenoleukodystrophy related; adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; ALDR; ALDRP; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2
Rabbit
Antigen affinity chromatography
RUO
225
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.77 mg/mL
0.1M Tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9UBJ2
ABCD2
Recombinant fragment corresponding to a region within amino acids 338 and 588 of ABCD2 (Uniprot ID#Q9UBJ2).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.