Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $511.50

Specifications

Antigen ABCD2
Concentration 1 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5118971
View Documents
Invitrogen™
PA5118971
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5122297
View Documents
Invitrogen™
PA5122297
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

ABCD2 Polyclonal Antibody for Western Blot, IHC (P)

ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.
Specifications

Specifications

ABCD2
-20°C
Polyclonal
Liquid
IgG
Mouse
ABCD2
ABC39; Abcd2; adrenoleukodystrophy related; adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; ALDR; ALDRP; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2
ABCD2
Primary
Antigen affinity chromatography
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
Q61285
26874
Recombinant protein ATP Binding Cassette Transporter D2. The antigen corresponds to amino acid range 501-695 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.