Learn More
Abnova™ ABCF3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055324T01
Catalog No. 89-932-164
Add to cart
Description
Description
Specifications
Specifications
293T
BC009253
pCMV-ABCF3 full-length
100 μL
Human
ABCF3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 79.7kDa
Transient overexpression cell lysate was tested with anti-ABCF3 antibody (Mfr. No. H00055324-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot