Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCF3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550288
Description
The antibody detects endogenous levels of total ABCF3 protein.
ABCF3 Polyclonal antibody specifically detects ABCF3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ABCF3 | |
Polyclonal | |
Unconjugated | |
ABCF3 | |
ABCF3; AI326318; ATP binding cassette subfamily F member 3; ATP-binding cassette sub-family F member 3; ATP-binding cassette, subfamily F (GCN20), member 3; ATP-binding cassette, sub-family F (GCN20), member 3; AU016058; BB119416; EST201864 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55324 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NUQ8 | |
ABCF3 | |
Fusion protein corresponding to a region derived from internal residues of human ATP-binding cassette, sub-family F (GCN20), member 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction