Learn More
ABCG2/CD338 Mouse anti-Human, Alexa Fluor 647, Clone: 5D3, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB995R100UG
Catalog No. FAB995R100
Add to cart
This item is not returnable. View return policy
Description
Description
ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ABCG2/CD338
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ABCG2
3T3 cells transduced with human ABCG2
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human ABCG2 in flow cytometry and immunocytochemistry.
Flow Cytometry
5D3
Flow Cytometry 0.25-1 ug/10^6 cells
ABC transporter, ABC15, ABCP, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXR, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Mouse
IgG2b
RUO
Primary
9429
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)