Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG2 Rabbit anti-Human, Unconjugated, Clone: AODG-1 , Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19516
Description
ABCG2 Monoclonal antibody specifically detects ABCG2 in Human samples. It is validated for WB-Ce.Specifications
ABCG2 | |
Monoclonal | |
Unconjugated | |
ABCG2 | |
Rabbit | |
100 μg | |
9429 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
AODG-1 | |
ATP-binding cassette, sub-family G (WHITE), member 2 | |
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, MGC102821, MRX, MXR, MXR1 | |
A synthetic peptide corresponding to human ABCG2 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction