Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550289

Catalog No. PA550289


Add to cart

Description

Description

The antibody detects endogenous levels of total ABCG4 protein.

May be involved in macrophage lipid homeostasis.
Specifications

Specifications

ABCG4
Polyclonal
Unconjugated
ABCG4
Liquid
ABCG4
Fusion protein corresponding to residues near the N terminal of human ATP-binding cassette, sub-family G (WHITE), member 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9H172
WHITE2
Rabbit
IgG
100 μL
-20°C
Primary
192663, 64137
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit