Learn More
Thermo Scientific™ Thermo-Fast PCR Plate, 24-well
Optimize PCR and qPCR applications with these 24-well semi-skirted plates.
Specifications
Description Semi-Skirted
No. of Wells 24
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
AB-0624B
View Documents
Thermo Scientific™
AB0624B
Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
Highlights
  • Conveniently pre-cut into 24-well segments
  • Suitable for use in 0.2 mL thermal cycler blocks
  • Semi-skirt adds rigidity and allows for labeling or barcoding
  • 0.3 mL maximum well volume (when used with adhesive seals)
  • Sealable using adhesive films and foils and all cap strips
Specifications
Specifications
Semi-Skirted
24