Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABH1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI4C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271390AF700

Catalog No. N271390A700


Add to cart

Description

Description

ABH1 Monoclonal antibody specifically detects ABH1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ABH1
OTI4C4
Immunohistochemistry
Affinity Purified, Conjugated
ALKBH1
Recombinant protein fragment corresponding to amino acids 1-245 of human ABH1 (NP_006011) produced in E.coli.
Immunogen affinity purified
DNA Repair
Primary
8846.0
Immunohistochemistry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit