Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179909

 View more versions of this product

Catalog No. NBP179909


Add to cart

Description

Description

ABH1 Polyclonal antibody specifically detects ABH1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

ABH1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ALKBH1
The specific Immunogen is proprietary information. Peptide sequence PGFIFIPNPFLPGYQWHWVKQCLKLYSQKPNVCNLDKHMSKEETQDLWEQ.
33 kDa
100 ul
DNA Repair
Primary
8846
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_006011
ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Rat: 86%; Mouse: 86%; Rabbit: 86%; Guinea pig: 86%; Horse: 79%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit