Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546200
Description
Peptide sequence: CIRFDYSGVG SSDGNSEEST LGKWRKDVLS IIDDLADGPQ ILVGSSLGGW Sequence homology: Human: 100%; Rabbit: 86%.
This gene encodes a mitochondrially-localized enzyme that acts in liver cells as a hydrolase. The encoded protein removes glucuronide from mycophenolic acid acyl-glucuronide. There is a pseudogene for this gene on chromosome 6. Alternative splicing results in multiple transcript variants.
Specifications
ABHD10 | |
Polyclonal | |
Unconjugated | |
ABHD10 | |
Abhd10; abhydrolase domain containing 10; abhydrolase domain containing 10, depalmitoylase; Abhydrolase domain-containing protein 10; abhydrolase domain-containing protein 10, mitochondrial; Alpha/beta hydrolase domain-containing protein 10; alpha/beta hydrolase domain-containing protein 10, mitochondrial; mycophenolic acid acyl-glucuronide esterase, mitochondrial; RGD1308084 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55347 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NUJ1 | |
ABHD10 | |
Synthetic peptide directed towards the middle region of human ABHD10. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction