Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD14A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101395
Description
Antibody detects endogenous levels of total ABHD14A.
ABHD14A play a possible role in granule neuron development.
Specifications
ABHD14A | |
Polyclonal | |
Unconjugated | |
ABHD14A | |
1110013B16Rik; ABHD14A; abhydrolase domain containing 14A; abhydrolase domain-containing protein 14A; abhydrolase domain-containing protein 14A; protein ABHD14A; alpha/beta hydrolase domain-containing protein 14A; AW558221; DORZ1; down-regulated in Zic1 deficient cerebellar primordium; downregulated in Zic1 deficient cerebellum; down-regulated in Zic-1-mutant protein; protein ABHD14A; RGD1309721; UNQ1913/PRO4373 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25864, 300982, 68644 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5I0C4, Q922Q6, Q9BUJ0 | |
ABHD14A | |
A synthesized peptide derived from human ABHD14A(Accession Q9BUJ0), corresponding to amino acid residues T109-D159. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction