Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHEB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551328

Catalog No. PA551328


Add to cart

Description

Description

The antibody detects endogenous levels of total ABHD14B protein.

ABHD14B is a 210 amino acid protein that localizes to both the cytoplasm and the nucleus where it exists as two alternatively spliced isoforms. Has hydrolase activity towards p-nitrophenyl butyrate (in vitro). May activate transcription.
Specifications

Specifications

ABHEB
Polyclonal
Unconjugated
ABHD14B
Liquid
ABHD14B
Full length fusion protein of human ABHD14B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q96IU4
CIB
Rabbit
IgG
100 μL
-20°C
Primary
84836
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit