Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD14B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 209521AP150UL
Description
ABHEB has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription.Specifications
ABHD14B | |
Polyclonal | |
Unconjugated | |
ABHD14B | |
ABHD14B, CCG1 interacting factor B, CIB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300983, 76491, 84836 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6DGG1, Q8VCR7, Q96IU4 | |
ABHD14B | |
ABHD14B Fusion Protein Ag15097 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction