Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569961
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
ABHD15 gene ontology annotations related to this gene include cellular lipid metabolic process.
Specifications
ABHD15 | |
Polyclonal | |
Unconjugated | |
ABHD15 | |
1300007F04Rik; ABHD15; abhydrolase domain containing 15; abhydrolase domain-containing protein 15; abhydrolase domain-containing protein 15; protein ABHD15; alpha/beta hydrolase; alpha/beta hydrolase domain-containing protein 15; alpha/beta hydrolase-1; lung alpha/beta hydrolase 1; protein ABHD15; RGD1304598; UNQ6510/PRO21435 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116236 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UXT9 | |
ABHD15 | |
synthetic peptide directed towards the N terminal region of human ABHD15. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction