Learn More
Abnova™ ABHD5 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L021T6
Catalog No. 89-932-193
Add to cart
Description
Description
Specifications
Specifications
HEK293
BC021958
Theoretical M.W. 39kDa
Transient overexpression cell lysate was tested with anti-ABHD5 antibody (Mfr. No. H00051099-M01) by Western Blot.
In modified RIPA Lysis Buffer.
2 mg/mL
ABHD5 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-ABHD5 full length
100 μg
Human
Immunoprecipitation, Western Blot