Learn More
Aviva Systems Biology Corp ABHD7 Antibody - N-terminal region (ARP55651_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABHD7. Alternate names: EPHXRP, FLJ90341, ABHD7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61566