Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHEB Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549963

Catalog No. PA549963


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ABHD14B protein.

ABHEB has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABHEB
Polyclonal
Unconjugated
ABHD14B
1810013B01Rik; ABHD14B; abhydrolase domain containing 14B; abhydrolase domain-containing protein 14B; Alpha/beta hydrolase domain-containing protein 14B; CCG1-interacting factor B; cell cycle gene 1-interacting factor B; CIB; epididymis secretory protein Li 299; HEL-S-299; Protein ABHD14B
Rabbit
Antigen affinity chromatography
RUO
76491, 84836
-20°C
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8VCR7, Q96IU4
ABHD14B
Synthesized peptide derived from internal of human ABHD14B (aa 161-210).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.