Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABI1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP257842

Catalog No. NBP257842


Add to cart

Description

Description

ABI1 Polyclonal antibody specifically detects ABI1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ABI1
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Abelson interactor 1, ABI-1, abl interactor 1, Abl-binding protein 4, AblBP4, abl-interactor 1, Abl-interactor protein 1 long, E3B1, eps8 binding protein, Eps8 SH3 domain-binding protein, Eps8-binding protein, interactor protein AblBP4, nap1 binding protein, Nap1-binding protein, Nap1BP, spectrin SH3 domain binding protein 1, Spectrin SH3 domain-binding protein 1, SSH3BP, SSH3BP1
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
ABI1
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SQYGTMTRQISRHNSTTSSTSSGGYRRTPSVTAQFSAQPHVNGGPLYSQ
Affinity purified
RUO
Primary
10006
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only