Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABI2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548398

Catalog No. PA548398

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ABI2 may act in regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. ABI2 is a part of the WAVE complex that regulates lamellipodia formation. The WAVE complex regulates actin filament reorganization via its interaction with the Arp2/3 complex. It regulates ABL1/c-Abl-mediated phosphorylation of MENA.
Specifications

Specifications

ABI2
Polyclonal
PBS with 0.09% sodium azide
Q9NYB9, P62484
Abl interactor 2, Abelson interactor 2, Abi-2, Abl-binding protein 3, AblBP3, Arg-binding protein 1, ArgBP1, ABI2, ARGBPIA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10152, 329165
Flow Cytometry, Western Blot
Unconjugated
ABI2
Liquid
ABI2
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ABI2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit