Learn More
ABI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148901AP
Catalog No. 50-172-9810
Add to cart
Description
Description
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.
Specifications
Specifications
ABI2
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62484, Q9NYB9
Abelson interactor 2, ABI 2, ABI2, ABI2B, Abl binding protein 3, abl interactor 2, AblBP3, AIP 1, Arg binding protein 1, ArgBP1, argBPIA, argBPIB, SSH3BP2
Rabbit
IgG
150 μL
-20°C
Primary
10152, 286928, 329165
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ABI2
Liquid
Abi2
ABI2 Fusion Protein Ag6679
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat