Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABL1 (phospho Y393) Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30835
Description
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1.
- High specificity
- High reactivity
Specifications
ABL1 (phospho Y393) | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1. | |
In PBS (0.09% sodium azide) | |
ABL/JTK7/bcr/abl/c-ABL/p150/v-abl | |
Rabbit | |
Protein A purification | |
RUO | |
25 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Dot Blot (1:100-500) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
P00519 | |
ABL1 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y393 of human ABL1. | |
400 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction