Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abl2 Rabbit anti-Human, Human, Mouse, Rat, Clone: ARC2200, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA537932

Encompass

Catalog No. PIMA537932


Add to cart

Description

Description

Positive Samples: HeLa, 293T, U-87MG, Rat testis

Abl2 Recombinant Monoclonal antibody specifically detects Abl2 in Human, Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Abl2
Recombinant Monoclonal
0.47 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P42684, Q4JIM5
AA536808; Abelson murine leukemia viral (v-abl) oncogene homolog 2; abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; ABL proto-oncogene 2, non-receptor tyrosine kinase; Abl1-related gene product; abl2; ABLL; Arg; arg tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; FLJ22224; FLJ31718; FLJ41441; tyrosine kinase ARG; Tyrosine-protein kinase ABL2; tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene 2; v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene); v-abl Abelson murine leukemia viral oncogene homolog 2; v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene)
A synthesized peptide derived from human ABL2
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Western Blot
ARC2200
Unconjugated
ABL2
Liquid
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11352, 27, 304883
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit