Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABLIM1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550292

Catalog No. PA550292


Add to cart

Description

Description

The antibody detects endogenous levels of total ABLIM1 protein.

This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

ABLIM1
Polyclonal
Unconjugated
ABLIM1
Liquid
ABLIM1
Fusion protein corresponding to a region derived from internal residues of human Actin binding LIM protein 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O14639, Q8K4G5
ABLIM, KIAA0059, LIMAB1
Rabbit
IgG
100 μL
-20°C
Primary
226251, 3983
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit