Learn More
GVS Maine ABLUO™ 13mm Syringe Filters, Sterile
Quick and efficient filtration of samples and all kind of solutions, solvents or gases
Manufacturer:  GVS Maine FJ13BSCPS002AL01
 View more versions of this product
Catalog No. 09-977-746
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Acrylic
FLL
Transparent
0.6cm2
MLL-MLS
Polyethersulfone (PES)