GVS Maine ABLUO™ 25mm Syringe Filters, Sterile
Filter devices designed to provide fast and efficient filtration of aqueous and organic solutions
Manufacturer: GVS Maine FJ25BSCPS002AL01
 View more versions of this product
Catalog No. 09-977-755
  Check Availability
Add to Cart
    Specifications
    Filtration Area (Metric) 4.6cm2
    Outlet Style MLL-MLS
    Color Transparent
    Housing Material Acrylic
    Inlet Style FLL
    Membrane Polyethersulfone (PES)
  • Description & Specifications
Related Products
Product Content Feedback