Learn More
Abnova™ 3-Color FISH Probe (chromosome 13, 18 and 21)
Labeled FISH probes for identification of prenatal chromosomal abberations using Fluorescent In Situ Hybridization Technique.
Manufacturer:  Abnova™ FM0001
Catalog No. 89-029-177
Add to cart
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
R6G, Texas Red, FITC
13q12.11, 18q11.2, 21q22.13
3-Color FISH Probe (chromosome 13, 18 & 21)
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell)
CEN13q, CEN18q, CEN21q
100 uL