Learn More
Abnova™ 3-Color FISH Probe (chromosome 21, X and Y)
Manufacturer:  Abnova™ FM0002
 View more versions of this product
Catalog No. 89-029-178
Add to cart
Description
Description
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
R6G, Texas Red, FITC
21q22.13, Xp11.23, Yq11.221
3-Color FISH Probe (chromosome 21, X & Y)
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell)
CEN21q, CENXp, CENYq
100 uL