Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Acetyl-CoA Assay Kit
To determine acetyl CoA level in a variety of samples, functional study
Supplier: Abnova™ KA0803
Specifications
10 to 100pM | |
Acetyl-CoA Assay Kit is used for determining acetyl CoA level in a variety of samples. | |
Functional Study | |
1 Kit | |
Activity Detection Assay Kit |
Store the kit at -20°C. | |
Fluorometric | |
25mL Acetyl CoA assay buffer, 0.2mL PicoProbe, 0.1mL conversion enzyme, 0.5mL Acetyl CoA enzyme mix, Acetyl CoA substrate mix (lyophilized), 1mL CoA quencher, quench remover (lyophilized), Acetyl CoA standard (1μM) (lyophilized) | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction