Learn More
Abnova™ Acid Phosphatase Assay Kit (Colorimetric)
To detect acid phosphatase (AP) activity, functional study
Manufacturer:  Abnova™ KA0816
Catalog No. 89-028-083
Add to cart
Specifications
Specifications
1 to 1000μM
Functional Study
1 Kit
Store the kit at -20°C.
Acid Phosphatase Assay Kit
Acid Phosphatase Assay Kit (Colorimetric) used for detecting AP activity.
100mL AP assay buffer, 1 vial pNPP substrate (10 TAB), 1 vial AP enzyme, 10mL stop solution
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric