Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Aconitase Assay Kit
Used for measuring aconitase activity and functional study
Supplier: Abnova™ KA0876
Specifications
Store the kit at 4°C. | |
Colorimetric | |
30mL assay buffer, 1 vial substrate, 1 vial Developer, 200μL enzyme mix, 1 vial Cysteine, 1 vial (NH4)Fe(SO4)2, 1 vial Isocitrate standard | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine, Food Sample, Fermentation Medium |
Aconitase Assay Kit is used for measuring aconitase activity. | |
Functional Study | |
1 Kit | |
Activity Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction