Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ACPA ELISA Kit
Indirect solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against citrullinated Proteins (ACP), in Human Serum or Plasma
Supplier: Abnova™ KA1268
Specifications
HRP | |
ACPA is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against citrullinated Proteins (ACP), in human serum or plasma. | |
Colorimetric | |
1 Kit | |
100 μL | |
Quantification |
Human | |
1U/mL | |
6 Vials, 1.5mL each combined calibrator with IgG class Anti-citrullinated protein Antibodies, 2 vials, 1.5mL each ACPA controls, 1 vial, 20mL Sample Buffer, 1 vial, 15mL Enzyme Conjugate solution, 1 vial, 15mL TMB Substrate solution, 1 plate 96-well plate, 1 vial, 20mL wash solution | |
Plasma, Serum | |
Store the kit at 4°C | |
ACPA ELISA kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction