Abnova™ ACPP (Human) ELISA Kit

Manufacturer: Abnova Corporation KA3185

 View more versions of this product

Catalog No. 89-100-887

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Cross Reactivity Human
    Detection Method Colorimetric
    For Use With (Application) Enzyme immunoassay for the quantitative measurement of ACPP in Serum
    Includes 1 x 96-well microtiter strip, 1 x 11mL sample diluent or Zero standard, 1 x 10mL Wash Buffer concentrate, 1 x 11mL Enzyme Conjugate, 0.70mL/each reference standard set, 1 x 11mL TMB solution, 1 x 11mL stop solution
    Product Type ACPP (Human) ELISA kit
    Quantity 96 Assays
    Sample Type Human Serum and Plasma
    Storage Requirements 4°C
    Sufficient For 1 x 96 wells
    Volume (Metric) Sample 50μL
    Conjugate HRP

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit