Learn More
Abnova™ ACPP (Human) ELISA Kit
Enzyme Immunoassay for the quantitative measurement of ACPP in Serum
Manufacturer:  Abnova™ KA1911
 View more versions of this product
Catalog No. 89-028-723
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
HRP
ACPP (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of ACPP in serum.
Quantification
ACPP (Human) ELISA Kit
1 Kit
Store the kit at 4°C.
25μL
Human
Colorimetric
1 x 96-well microtiter strip, 1 x 11mL sample diluent or Zero standard, 1 x 10mL Wash Buffer concentrate, 1 x 11mL Enzyme Conjugate, 0.70mL/each reference standard set, 1 x 11mL TMB solution, 1 x 11mL stop solution
ACPP (Human) ELISA kit
Serum
1 x 96 wells