Learn More
Abnova™ Adipogenesis Assay Kit
Used for quantifying triglyceride accumulation in cells and tissues and functional study
Manufacturer:  Abnova™ KA0902
Catalog No. 89-024-451
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
0.2 to 10nm
Functional Study
1 Kit
Store the kit at -20°C.
Adipogenesis Assay Kit
Adipogenesis Assay Kit is used for quantifying triglyceride accumulation in cells and tissues.
25mL Adipogenesis assay buffer, 10mL Lipid extraction solution, 200μL Adipogenesis probe (in DMSO solution), 1 vial Lipase (lyophilized), 1 vial Adipogenesis enzyme mix (lyophilized), 0.3mL Triglyceride standard (1mM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric