Learn More
Abnova™ ADP Assay Kit (Colorimetric/Fluorometric)
To provide a convenient colorimetric and fluorometric means to measure ADP level, functional study
Manufacturer:  Abnova™ KA0807
Catalog No. 89-028-091
Add to cart
Specifications
Specifications
ADP Assay Kit (Colorimetric/Fluorometric) is used for providing a convenient colorimetric and fluorometric means to measure ADP level.
25mL ADP assay buffer, 200μL ADP probe, 1 vial ADP convertor, 1 vial ADP developer mix, 1 vial ADP standard
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
ADP Assay Kit