Learn More
Abnova™ Aggrecanase Activity Assay Kit
Colorimetric method for assaying the activity of aggrecanase
Manufacturer:  Abnova™ KA1497
Catalog No. 89-028-282
Add to cart
Specifications
Specifications
Aggrecanase Activity Assay Kit is a colorimetric method for assaying the activity of aggrecanase.
Aggrecan-IGD, ADAMTS4-standard, inhibitor solution, reaction buffer, EDTA Dilution buffer, microtiter plate, ELISA buffer, ARGSVIL-peptide standard, antibody-peroxidase conjugate, wash buffer, detection solution TMB, Sulfuric Acid
100μL
Colorimetric
Functional Study
1 Kit
Store Aggrecan-IGD, Inhibitor Solution, ADAMTS4-Standard, ARGSVIL-Peptide Standard at -20°C. Store all other components at 4°C.
Aggrecanase Activity Assay Kit