Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Alkaline Phosphatase Assay Kit (Fluorometric)
To detect alkaline phosphatase (ALP) activity, functional study
Supplier: Abnova™ KA0820
Includes:
- ALP assay buffer, 100mL
- MUP substrate, 1 vial
- ALP enzyme, 1 vial
- Stop solution, 25mL
Specifications
Store the kit at -20°C. | |
Fluorometric | |
100mL ALP assay buffer, 1 vial MUP substrate, 1 vial ALP enzyme, 25mL stop solution | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Alkaline Phosphatase Assay Kit (Fluorometric) is used for detecting ALP activity. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction