Learn More
Abnova™ Alpha Fodrin Ab IgG/IgA ELISA Kit
indirect solid phase ELISA for the quantitative measurement of IgG and IgA class autoantibodies against Alpha-fodrin in Human Serum or Plasma
Manufacturer:  Abnova™ KA1087
Catalog No. 89-027-225
Add to cart
Specifications
Specifications
HRP
Colorimetric
6 Vials, 1.5mL each combined calibrator with IgG and IgA class Anti-alpha-fodrin Antibodies, 2 vials, 1.5mL each Anti-alpha-fodrin controls, 1 vial 20mL Sample Buffer, 1 vial 15mL Enzyme Conjugate solution, 1 vial 15mL TMB Substrate solution, 1 vial, 15mL stop solution, 1 plate 96-well plate, 1 vial, 20mL wash solution
96 Tests
Store the kit at 4°C.
Alpha Fodrin IgG/IgA ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG and IgA class autoantibodies against alpha-Fodrin in human serum or plasma.
Quantification
Alpha Fodrin Ab IgG/IgA ELISA Kit
Plasma, Serum
10μL