Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ AMA-M2 Ab ELISA Kit

Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against mitochondrial M2 subtype antigen in Human Serum or Plasma

Manufacturer:  Abnova™ KA1078

Catalog No. 89-027-216


Add to cart

Includes:
  • Divisible microplate consisting of 12 modules of 8 wells coated with highly purified mitochondrial M2 subtype antigen. Ready to use, Qty.1
  • AMA-M2 Calibrators (A-F) in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) containing: IgG: 0; 12.5; 25; 50; 100; and 200 IU/mL, Ready to use, 6 vials, 1.5mL each
  • AMA-M2 Controls in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) positive (1) and negative (2), for the respective concentrations see the enclosed package insert. Ready to use, 2 vials, 1, 5mL each
  • Sample buffer (Tris, NaN3 <0.1% (w/w)), yellow, concentrate (5x) 1 vial, 20mL
  • Enzyme conjugate solution (PBS, PROCLIN 300 <0.5% (v/v)), (light red) containing polyclonal rabbit anti-human IgG; labelled with horseradish peroxidase. Ready to use, 1 vial, 15mL
  • TMB substrate solution. Ready to use, 1 vial, 15mL

Specifications

Specifications

HRP
Colorimetric
6 Vials, 1.5mL each AMA-M2 calibrator (A-F) in a Serum/Buffer Matrix, 2 vials, 1,5mL each AMA-M2 controls in a Serum/Buffer Matrix, 1 vial, 20mL Sample Buffer, 1 vial, 15mL Enzyme Conjugate solution containing Polyclonal Rabbit Anti-Human IgG, 1 vial 15mL TMB Substrate solution, 1 vial 15mL stop solution, 1 vial, 20mL wash solution, 1 plate 96-well microplate
96 Assays
Store the kit at 4°C.
AMA-M2 Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against mitochondrial M2 subtype antigen in human serum or plasma.
Quantification
AMA-M2 Ab ELISA Kit
Plasma, Serum
10μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit