Learn More
Abnova™ Ammonia Assay Kit
Used for detecting ammonia
Manufacturer:  Abnova™ KA0810
Catalog No. 89-028-088
Add to cart
Description
Description
  • In the assay, Ammonia or ammonium is converted to a product that reacts with the OxiRed probe to generate color (OD 570nm) which can be easily quantified by plate reader
  • The kit can detect 1 nmol (≈20 μM) of ammonia or ammonium, much more sensitive than measuring NADPH-based ammonia assay
Specifications
Specifications
Ammonia Assay Kit is used for detecting ammonia.
25mL ammonia assay buffer, 200μL OxiRed™ probe in DMSO, 1 vial enzyme mix (lyophilized), 1 vial developer, 1 vial Converting enzyme (lyophilized), 100μL NH4Cl standard (10mM)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric
Functional Study
1 Kit
Store the kit at -20°C.
Ammonia Assay Kit