Learn More
Abnova™ Amphetamine ELISA Kit
Qualitative, specific and sensitive in-vitro test to detect the presence of d-amphetamine in samples such as whole Blood, Oral fluids, Serum, Plasma and Urine
Manufacturer:  Abnova™ KA0926
Catalog No. 89-024-467
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Colorimetric
12 x 8 x 1 Microwell strip, 12.5mL Enzyme Conjugate, 1mL positive reference standard, 1mL negative control, 14mL TMB chromogenic Substrate, 12.5mL stop solution
96 Assays
Store the kit at 4°C.
Amphetamine ELISA Kit is a specific and sensitive in-vitro test to detect the presence of d-amphetamine in samples such as whole blood, oral fluids, serum, plasma and urine.
Qualitative
Amphetamine ELISA Kit
Oral Fluid, Plasma, Serum, Urine, Whole Blood
10μL