Learn More
Abnova™ ANGPT1 (Human) ELISA Kit
In-Vitro ELISA for the quantitative measurement of Human ANGPT1
Manufacturer:  Abnova™ KA1866
Catalog No. 89-028-577
Add to cart
Specifications
Specifications
Biotin
ANGPT1 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human ANGPT1.
Quantification
ANGPT1 (Human) ELISA Kit
Cell Culture Supernatant, Plasma (Heparin), Serum, Urine
100μL
Human
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP-streptavidian concentrate, 12mL TMB one-step Substrate reagent, 8mL stop solut
96 Assays
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.